2,4-Difluorobenzhydrazide 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Difluorobenzoic acid hydrazide, 2,4-Difluorobenzohydrazide, 2,4-Difluorobenzoic hydrazide, 2,4-Difluorobenzene-1-carbohydrazide
PC2664-1G PC2664-5G
APOSPC2664-1GEA 21.9 EUR
APOSPC2664-1G APOSPC2664-5G
2,4-Difluorobenzhydrazide 98%
2,4-Difluorobenzhydrazide
Formula: C₇H₆F₂N₂O
Melting Pt: 141…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00173685
CAS Number: 118737-62-5