4-Iodo-2-(trifluoromethyl)benzonitrile
Supplier: Apollo Scientific
Warning
PC2697-25G PC2697-5G PC2697-100G
APOSPC2697-25GEA 49.3 EUR
APOSPC2697-25G APOSPC2697-5G APOSPC2697-100G
4-Iodo-2-(trifluoromethyl)benzonitrile
4-Iodo-2-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₃IN
MW: 297.02 g/mol
Melting Pt: 50…55 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD08458091
CAS Number: 101066-87-9
UN: 3439
ADR: 6.1 III