2,3-Difluorobenzyl bromide 98%
Supplier: Apollo Scientific
Danger
Synonyms: α-Bromo-2,3-difluorotoluene
PC2739-100G PC2739-25G PC2739-5G
APOSPC2739-100GEA 249 EUR
APOSPC2739-100G APOSPC2739-25G APOSPC2739-5G
2,3-Difluorobenzyl bromide 98%
2,3-Difluorobenzyl bromide
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 186
Melting Pt: 8
Density: 1.628 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00042488
CAS Number: 113211-94-2
UN: 3265
ADR: 8 II