α-Bromo-2,5-difluorotoluene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Bromomethyl)-1,4-difluorobenzene, 2,5-Difluorobenzyl bromide
PC2742-10G PC2742-250G PC2742-50G
APOSPC2742-10GEA 21.9 EUR
APOSPC2742-10G APOSPC2742-250G APOSPC2742-50G
α-Bromo-2,5-difluorotoluene 98%
α-Bromo-2,5-difluorotoluene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 186
Melting Pt: 8
Density: 1.609 g/cm³ (25 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00009897
CAS Number: 85117-99-3
UN: 2924
ADR: 3+8 II