2-Amino-4-(trifluoromethoxy)phenol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-5-(trifluoromethoxy)aniline
PC2780-5G PC2780-1G
APOSPC2780-5GEA 313 EUR
APOSPC2780-5G APOSPC2780-1G
2-Amino-4-(trifluoromethoxy)phenol 98%
2-Amino-4-(trifluoromethoxy)phenol
Formula: C₇H₆F₃NO₂
MW: 193.13 g/mol
Melting Pt: 98…101 °C
Storage Temperature: Refrigerator
CAS Number: 461699-34-3