2-Fluoro-3-nitrotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-1-methyl-3-nitrobenzene
PC2851-100G PC2851-25G
APOSPC2851-100GEA 116 EUR
APOSPC2851-100G APOSPC2851-25G
2-Fluoro-3-nitrotoluene
2-Fluoro-3-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 110…111 °C/12mmHg
Melting Pt: 18…19 °C
Density: 1.28 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03412242
CAS Number: 437-86-5