2-Amino-6-(trifluoromethyl)benzonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-6-trifluoromethylbenzonitrile
PC2853-1G PC2853-250MG PC2853-5G
APOSPC2853-1GEA 27.7 EUR
APOSPC2853-1G APOSPC2853-250MG APOSPC2853-5G
2-Amino-6-(trifluoromethyl)benzonitrile
2-Amino-6-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.1339 g/mol
Melting Pt: 70…73 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08741381
CAS Number: 58458-11-0
UN: 3439
ADR: 6.1 III