(2,6-Difluorophenyl)acetonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(2,6-Difluorophenyl)acetonitrile, 2,6-Difluorobenzyl cyanide
PC2874EJ-25G PC2874EJ-5G PC2874EJ-100G
APOSPC2874EJ-25GEA 43.5 EUR
APOSPC2874EJ-25G APOSPC2874EJ-5G APOSPC2874EJ-100G
(2,6-Difluorophenyl)acetonitrile 98%
(2,6-Difluorophenyl)acetonitrile
Formula: C₈H₅F₂N
Boiling Pt: 109 °C/27mmHg
Density: 1.238 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00009973
CAS Number: 654-01-3
EINECS: 211-504-7
UN: 1993
ADR: 3,III