(3,4-Difluorophenyl)acetonitrile 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Difluorobenzyl cyanide, 2-(3,4-Difluorophenyl)acetonitrile
PC2874EK-100G PC2874EK-25G PC2874EK-5G
APOSPC2874EK-100GEA 0 EUR
APOSPC2874EK-100G APOSPC2874EK-25G APOSPC2874EK-5G
(3,4-Difluorophenyl)acetonitrile 97%
(3,4-Difluorophenyl)acetonitrile
Formula: C₈H₅F₂N
MW: 153.13 g/mol
Boiling Pt: 110 °C/13mmHg
Density: 1.244 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009974
CAS Number: 658-99-1
EINECS: 211-528-8
UN: 3276
ADR: 6.1 III