1-(2,4-Difluorophenyl)-2-thiourea 97%
Supplier: Apollo Scientific
Danger
Synonyms: N-(2,4-Difluorophenyl)thiourea, 1-(2,4-Difluorophenyl)thiourea, 2,4-Difluorophenylthiourea, (2,4-Difluorophenyl)thiourea
PC2876-1G PC2876-25G PC2876-5G
APOSPC2876-1GEA 21.9 EUR
APOSPC2876-1G APOSPC2876-25G APOSPC2876-5G
1-(2,4-Difluorophenyl)-2-thiourea 97%
1-(2,4-Difluorophenyl)-2-thiourea
Formula: (C₆H₃F₂)NC(S)NH₂
Melting Pt: 164…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00041150
CAS Number: 175277-76-6
UN: 2811
ADR: 6.1,III