2,2-Difluoropropane 97%
Supplier: Apollo Scientific
Warning
PC2877E-25G PC2877E-5G
APOSPC2877E-25GEA 290 EUR
APOSPC2877E-25G APOSPC2877E-5G
2,2-Difluoropropane 97%
2,2-Difluoropropane
Formula: C₃H₆F₂
Boiling Pt: –1 °C
Density: 0.92 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 420-45-1
UN: 3161
ADR: 2.1,