1-(4-(Trifluoromethyl)pyrimidin-2-yl)piperidin-4-ol 95+%
Supplier: Apollo Scientific
PC2931-250MG PC2931-1G
APOSPC2931-250MGEA 99.2 EUR
APOSPC2931-250MG APOSPC2931-1G
1-(4-(Trifluoromethyl)pyrimidin-2-yl)piperidin-4-ol 95+%
1-(4-(Trifluoromethyl)pyrimidin-2-yl)piperidin-4-ol
Formula: C₁₀H₁₂F₃N₃O
MW: 247.2200 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD01311906
CAS Number: 401930-07-2