5-[3,5-Di(trifluoromethyl)phenyl]-2-furaldehyde 97%
Supplier: Apollo Scientific
Synonyms: 5-[3,5-Bis(trifluoromethyl)phenyl]furan-2-carboxaldehyde
PC2982-10G PC2982-1G PC2982-5G
APOSPC2982-10GEA 346 EUR
APOSPC2982-10G APOSPC2982-1G APOSPC2982-5G
5-[3,5-Di(trifluoromethyl)phenyl]-2-furaldehyde 97%
5-[3,5-Di(trifluoromethyl)phenyl]-2-furaldehyde
Formula: C₁₃H₆F₆O₂
MW: 308.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173833
CAS Number: 256658-04-5