6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Supplier: Apollo Scientific
Warning
PC300793-1G PC300793-250MG
APOSPC300793-1GEA 139 EUR
APOSPC300793-1G APOSPC300793-250MG
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Formula: C₉H₅F₃N₂O₂
MW: 230.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258865
CAS Number: 1018828-69-7