Dimethyl hexafluoroglutarate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Dimethyl perfluoroglutarate
PC3015M-100G PC3015M-25G
APOSPC3015M-100GEA 415 EUR
APOSPC3015M-100G APOSPC3015M-25G
Dimethyl hexafluoroglutarate 98%
Dimethyl hexafluoroglutarate
Formula: C₇H₆F₆O₄
Boiling Pt: 100 °C/34mmHg
Melting Pt: –32.7 °C
Density: 1.4856 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00155851
CAS Number: 1513-62-8