2,4-Dinitro-5-fluoroaniline 97%
Supplier: Apollo Scientific
Warning
PC3020-25G PC3020-5G
APOSPC3020-25GEA 277 EUR
APOSPC3020-25G APOSPC3020-5G
2,4-Dinitro-5-fluoroaniline 97%
2,4-Dinitro-5-fluoroaniline
Formula: C₆H₄FN₃O₄
Melting Pt: 185…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00007150
CAS Number: 367-81-7