2,3-Dichloro-6-fluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
PC302005-25G PC302005-5G PC302005-1G PC302005-100G
APOSPC302005-25GEA 346 EUR
APOSPC302005-25G APOSPC302005-5G APOSPC302005-1G APOSPC302005-100G
2,3-Dichloro-6-fluorobenzoic acid 97%
2,3-Dichloro-6-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₂
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00090506
CAS Number: 32890-91-8