2,3-Dichloro-6-fluorobenzylamine 97%
Supplier: Apollo Scientific
PC302006-1G PC302006-5G
APOSPC302006-1GEA 108 EUR
APOSPC302006-1G APOSPC302006-5G
2,3-Dichloro-6-fluorobenzylamine 97%
2,3-Dichloro-6-fluorobenzylamine
Formula: C₇H₆Cl₂FN
MW: 194.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660174
CAS Number: 886501-27-5