2,3-Dichloro-6-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
PC302067-1G PC302067-5G
APOSPC302067-1GEA 116 EUR
APOSPC302067-1G APOSPC302067-5G
2,3-Dichloro-6-(trifluoromethyl)benzonitrile 98%
2,3-Dichloro-6-(trifluoromethyl)benzonitrile
Formula: C₈H₂Cl₂F₃N
Storage Temperature: Ambient
MDL Number: MFCD09832276
CAS Number: 186517-39-5
UN: 3439
ADR: 6.1 III