(2,6-Dichloro-3-(trifluoromethyl)phenyl)acetonitrile 98%
Supplier: Apollo Scientific
PC302071-1G PC302071-5G
APOSPC302071-1GEA 116 EUR
APOSPC302071-1G APOSPC302071-5G
(2,6-Dichloro-3-(trifluoromethyl)phenyl)acetonitrile 98%
(2,6-Dichloro-3-(trifluoromethyl)phenyl)acetonitrile
Formula: C₉H₄Cl₂F₃N
Storage Temperature: Ambient
MDL Number: MFCD11519350
CAS Number: 1092461-09-0