3-Methoxy-2-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
Warning
PC302096-1G PC302096-5G PC302096-10G
APOSPC302096-1GEA 98.6 EUR
APOSPC302096-1G APOSPC302096-5G APOSPC302096-10G
3-Methoxy-2-(trifluoromethyl)benzonitrile 98%
3-Methoxy-2-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃NO
MW: 201.15 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD13185684
CAS Number: 1214385-02-0
UN: 3439
ADR: 6.1 III