3-[4-Fluoro-3-methyl-5-(trifluoromethyl)phenyl]propionic acid 97%
Supplier: Apollo Scientific
Synonyms: 3-(4-Fluoro-3-methyl-5-(trifluoromethyl)phenyl)propanoic acid
PC302190-1G PC302190-250MG
APOSPC302190-1GEA 371 EUR
APOSPC302190-1G APOSPC302190-250MG
3-[4-Fluoro-3-methyl-5-(trifluoromethyl)phenyl]propionic acid 97%
3-[4-Fluoro-3-methyl-5-(trifluoromethyl)phenyl]propionic acid
Formula: C₁₁H₁₀F₄O₂
MW: 250.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201076
CAS Number: 1373920-73-0