(2,3-Dichloro-6-(trifluoromethyl)phenyl)acetonitrile 98%
Supplier: Apollo Scientific
PC302221-1G PC302221-5G
APOSPC302221-1GEA 136 EUR
APOSPC302221-1G APOSPC302221-5G
(2,3-Dichloro-6-(trifluoromethyl)phenyl)acetonitrile 98%
(2,3-Dichloro-6-(trifluoromethyl)phenyl)acetonitrile
Formula: C₉H₄Cl₂F₃N
Melting Pt: 49…52 °C
Storage Temperature: Ambient
MDL Number: MFCD09832275
CAS Number: 1017777-89-7