2-Methoxy-3-(trifluoromethyl)benzylamine 98%
Supplier: Apollo Scientific
Danger
PC302246-1G PC302246-5G
APOSPC302246-1GEA 136 EUR
APOSPC302246-1G APOSPC302246-5G
2-Methoxy-3-(trifluoromethyl)benzylamine 98%
2-Methoxy-3-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃NO
MW: 205.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832318
CAS Number: 1017778-87-8
UN: 2735
ADR: 8 III