3-Chloro-5-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC302290-25G PC302290-5G PC302290-1G
APOSPC302290-25GEA 202 EUR
APOSPC302290-25G APOSPC302290-5G APOSPC302290-1G
3-Chloro-5-(trifluoromethyl)benzaldehyde 97%
3-Chloro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD04115845
CAS Number: 477535-43-6