4-Methoxy-2-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Danger
PC302343-1G PC302343-5G
APOSPC302343-1GEA 79.9 EUR
APOSPC302343-1G APOSPC302343-5G
4-Methoxy-2-(trifluoromethyl)phenylacetonitrile 98%
4-Methoxy-2-(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₈F₃NO
Melting Pt: 30…33 °C
Storage Temperature: Ambient
MDL Number: MFCD09025407
CAS Number: 916420-88-7
UN: 3439
ADR: 6.1 III