2,6-Dichloro-3-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC302378-25G PC302378-5G PC302378-1G
APOSPC302378-25GEA 579 EUR
APOSPC302378-25G APOSPC302378-5G APOSPC302378-1G
2,6-Dichloro-3-(trifluoromethyl)benzaldehyde 98%
2,6-Dichloro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₃Cl₂F₃O
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD11519344
CAS Number: 134741-65-4