(6-Chloro-2-fluoro-3-methoxyphenyl)acetic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Chloro-2-fluoro-3-methoxyphenylacetic acid
PC302634-1G PC302634-5G PC302634-10G
APOSPC302634-1GEA 60.9 EUR
APOSPC302634-1G APOSPC302634-5G APOSPC302634-10G
(6-Chloro-2-fluoro-3-methoxyphenyl)acetic acid 98%
(6-Chloro-2-fluoro-3-methoxyphenyl)acetic acid
Formula: C₉H₈ClFO₃
MW: 218.61 g/mol
Melting Pt: 134…137 °C
Storage Temperature: Ambient
MDL Number: MFCD09832273
CAS Number: 1017777-83-1