2-(Difluoromethoxy)-5-fluorobenzamide
Supplier: Apollo Scientific
Warning
PC302652-1G PC302652-5G
APOSPC302652-1GEA 136 EUR
APOSPC302652-1G APOSPC302652-5G
2-(Difluoromethoxy)-5-fluorobenzamide
2-(Difluoromethoxy)-5-fluorobenzamide
Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652418
CAS Number: 1240256-77-2