2-Methoxy-6-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Danger
PC302696-1G PC302696-5G
APOSPC302696-1GEA 212 EUR
APOSPC302696-1G APOSPC302696-5G
2-Methoxy-6-(trifluoromethyl)phenylacetonitrile 98%
2-Methoxy-6-(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₈F₃NO
Melting Pt: 48…51 °C
Storage Temperature: Ambient
MDL Number: MFCD09832327
CAS Number: 1017779-07-5
UN: 3439
ADR: 6.1,III