4-Ethoxy-2,3-difluorobenzylamine
Supplier: Apollo Scientific
PC302782-1G PC302782-5G
APOSPC302782-1GEA 171 EUR
APOSPC302782-1G APOSPC302782-5G
4-Ethoxy-2,3-difluorobenzylamine
4-Ethoxy-2,3-difluorobenzylamine
Formula: C₉H₁₁F₂NO
MW: 187.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258708
CAS Number: 1017779-64-4