2-Chloro-4,5-difluorobenzylamine 98%
Supplier: Apollo Scientific
Danger
Synonyms: (2-Chloro-4,5-difluorophenyl)methanamine
PC302793-5G PC302793-1G
APOSPC302793-5GEA 177 EUR
APOSPC302793-5G APOSPC302793-1G
2-Chloro-4,5-difluorobenzylamine 98%
2-Chloro-4,5-difluorobenzylamine
Formula: C₇H₆ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD06213035
CAS Number: 771582-19-5
UN: 2735
ADR: 8,II