4'-Ethoxy-3'-(trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
PC302872-5G PC302872-1G
APOSPC302872-5GEA 348 EUR
APOSPC302872-5G APOSPC302872-1G
4'-Ethoxy-3'-(trifluoromethyl)acetophenone 98%
4'-Ethoxy-3'-(trifluoromethyl)acetophenone
Formula: C₁₁H₁₁F₃O₂
MW: 232.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13196590
CAS Number: 851263-13-3