6-Chloro-2,3-difluorophenol 98%
Supplier: Apollo Scientific
Synonyms: 6-Chloro-2,3-difluorophenol
PC302898-10G PC302898-5G PC302898-25G
APOSPC302898-10GEA 294 EUR
APOSPC302898-10G APOSPC302898-5G APOSPC302898-25G
6-Chloro-2,3-difluorophenol 98%
6-Chloro-2,3-difluorophenol
Formula: C₆H₃ClF₂O
MW: 164.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201050
CAS Number: 186590-18-1