2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetic acid 98%
Supplier: Apollo Scientific
Synonyms: 2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetic acid
PC302922-1G PC302922-5G
APOSPC302922-1GEA 266 EUR
APOSPC302922-1G APOSPC302922-5G
2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetic acid 98%
2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetic acid
Formula: C₁₀H₈F₄O₂
MW: 236.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687200
CAS Number: 1323966-46-6