2'-Methyl-3'-(trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2-Methyl-3-(trifluoromethyl)phenyl)ethanone
PC302932-10G PC302932-5G PC302932-1G
APOSPC302932-10GEA 328 EUR
APOSPC302932-10G APOSPC302932-5G APOSPC302932-1G
2'-Methyl-3'-(trifluoromethyl)acetophenone 98%
2'-Methyl-3'-(trifluoromethyl)acetophenone
Formula: C₁₀H₉F₃O
MW: 202.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832283
CAS Number: 1017778-01-6