4'-Chloro-3'-(trifluoromethoxy)acetophenone 98%
Supplier: Apollo Scientific
Warning
PC302976-10G PC302976-5G PC302976-1G
APOSPC302976-10GEA 429 EUR
APOSPC302976-10G APOSPC302976-5G APOSPC302976-1G
4'-Chloro-3'-(trifluoromethoxy)acetophenone 98%
4'-Chloro-3'-(trifluoromethoxy)acetophenone
Formula: C₉H₆ClF₃O₂
Melting Pt: 40…43 °C
Storage Temperature: Ambient
MDL Number: MFCD06660263
CAS Number: 886501-62-8