6-Chloro-2,3-difluoroanisole 97%
Supplier: Apollo Scientific
PC303011-5G PC303011-1G
APOSPC303011-5GEA 192 EUR
APOSPC303011-5G APOSPC303011-1G
6-Chloro-2,3-difluoroanisole 97%
6-Chloro-2,3-difluoroanisole
Formula: C₇H₅ClF₂O
MW: 178.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201078
CAS Number: 1373921-04-0