2-Bromo-3,6-difluorobenzaldehyde
Supplier: Apollo Scientific
Warning
PC303025-10G PC303025-1G PC303025-5G PC303025-25G
APOSPC303025-10GEA 234 EUR
APOSPC303025-10G APOSPC303025-1G APOSPC303025-5G APOSPC303025-25G
2-Bromo-3,6-difluorobenzaldehyde
2-Bromo-3,6-difluorobenzaldehyde
Formula: C₇H₃BrF₂O
MW: 221.00 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12922618
CAS Number: 934987-26-5