2-Methoxy-6-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
Danger
PC303044-10G PC303044-5G PC303044-1G
APOSPC303044-10GEA 485 EUR
APOSPC303044-10G APOSPC303044-5G APOSPC303044-1G
2-Methoxy-6-(trifluoromethyl)benzonitrile 98%
2-Methoxy-6-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃NO
MW: 201.15 g/mol
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD09832322
CAS Number: 1017778-93-6
UN: 3439
ADR: 6.1 III