4-Methoxy-3-methyl-5-(trifluoromethyl)benzonitrile 97%
Supplier: Apollo Scientific
PC303130-5G PC303130-1G
APOSPC303130-5GEA 330 EUR
APOSPC303130-5G APOSPC303130-1G
4-Methoxy-3-methyl-5-(trifluoromethyl)benzonitrile 97%
4-Methoxy-3-methyl-5-(trifluoromethyl)benzonitrile
Formula: C₁₀H₈F₃NO
MW: 215.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22682821
CAS Number: 1431329-59-7
UN: 3439
ADR: 6.1 III