2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: Flecainide Impurity D
PC3053N-100G PC3053N-25G PC3053N-5G
APOSPC3053N-100GEA 46.4 EUR
APOSPC3053N-100G APOSPC3053N-25G APOSPC3053N-5G
2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid 97%
2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid
Formula: C₁₁H₈F₆O₄
MW: 318.17 g/mol
Melting Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00221424
CAS Number: 35480-52-5
EINECS: 609-132-9