2-Amino-5-(trifluoromethyl)benzonitrile
Supplier: Apollo Scientific
Danger
Synonyms: 4-Amino-3-cyanobenzotrifluoride, 2-Amino-5-trifluoromethylbenzonitrile
PC3059-1G PC3059-5G
APOSPC3059-1GEA 21.9 EUR
APOSPC3059-1G APOSPC3059-5G
2-Amino-5-(trifluoromethyl)benzonitrile
2-Amino-5-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.1300 g/mol
Boiling Pt: 95…115 °C/0.4mmHg
Melting Pt: 72…74 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03407465
CAS Number: 6526-08-5
UN: 3439
ADR: 6.1 III