2-Methoxy-4-(trifluoromethyl)phenylboronic acid
Supplier: Apollo Scientific
Synonyms: (2-Methoxy-4-(trifluoromethyl)phenyl)boronic acid
PC3077-5G PC3077-10G PC3077-1G
APOSPC3077-5GEA 93.4 EUR
APOSPC3077-5G APOSPC3077-10G APOSPC3077-1G
2-Methoxy-4-(trifluoromethyl)phenylboronic acid
2-Methoxy-4-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₃
MW: 219.9500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03428511
CAS Number: 312936-89-3