2-Bromo-6-fluoropyridine 97+%
Supplier: Apollo Scientific
Danger
PC3082-25G PC3082-5G PC3082-100G
APOSPC3082-25GEA 55.1 EUR
APOSPC3082-25G APOSPC3082-5G APOSPC3082-100G
2-Bromo-6-fluoropyridine 97+%
2-Bromo-6-fluoropyridine
Formula: C₅H₃BrFN
Boiling Pt: 162…164 °C
Melting Pt: 30…34 °C
Density: 1.71 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD03095258
CAS Number: 144100-07-2
UN: 2811
ADR: 6.1,III