4,4'-Bis(trifluoromethyl)-2,2'-dinitrodiphenyldisulphide 97%
Supplier: Apollo Scientific
PC3106M-5G PC3106M-25G
APOSPC3106M-5GEA 132 EUR
APOSPC3106M-5G APOSPC3106M-25G
4,4'-Bis(trifluoromethyl)-2,2'-dinitrodiphenyldisulphide 97%
4,4'-Bis(trifluoromethyl)-2,2'-dinitrodiphenyldisulphide
Formula: C₁₄H₆F₆N₂S₂O₄
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00018043
CAS Number: 860-39-9