1,6-Divinylperfluorohexane 99%
Supplier: Apollo Scientific
Warning
PC3120-1G PC3120-25G PC3120-5G PC3120-100G
APOSPC3120-1GEA 0 EUR
APOSPC3120-1G APOSPC3120-25G APOSPC3120-5G APOSPC3120-100G
1,6-Divinylperfluorohexane 99%
1,6-Divinylperfluorohexane
Formula: C₁₀H₆F₁₂
Boiling Pt: 83.5 °C/43mmHg
Density: 1.332 g/cm³ (25 °C)
Flash Pt: 67.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00077521
CAS Number: 1800-91-5