4-(Trifluoromethyl)pyridine-3-carboxamideoxime
Supplier: Apollo Scientific
|
Warning
|
PC3123-250MG
PC3123-1G
APOSPC3123-250MGEA
63.8
EUR
APOSPC3123-250MG
APOSPC3123-1G
4-(Trifluoromethyl)pyridine-3-carboxamideoxime
4-(Trifluoromethyl)pyridine-3-carboxamideoxime
Formula:
C₇H₆F₃N₃O Melting Pt: 183…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00219948 CAS Number: 175204-85-0 |