2-Fluoro-6-(4-(methylthio)phenoxy)benzonitrile
Supplier: Apollo Scientific
Danger
PC31842-250MG PC31842-1G
APOSPC31842-250MGEA 21.9 EUR
APOSPC31842-250MG APOSPC31842-1G
2-Fluoro-6-(4-(methylthio)phenoxy)benzonitrile
2-Fluoro-6-(4-(methylthio)phenoxy)benzonitrile
Formula: C₁₄H₁₀FNOS
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00068069
CAS Number: 148901-52-4
UN: 3439
ADR: 6.1,III