1-Ethyl-4-fluorobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-4-ethylbenzene, (4-Fluorophenyl)ethane, 4-Ethylfluorobenzene
PC3208-1G PC3208-25G PC3208-5G
APOSPC3208-1GEA 65.4 EUR
APOSPC3208-1G APOSPC3208-25G APOSPC3208-5G
1-Ethyl-4-fluorobenzene 97%
1-Ethyl-4-fluorobenzene
Formula: C₈H₉F
Boiling Pt: 139 °C
Density: 0.98 g/cm³ (25 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00039218
CAS Number: 459-47-2
UN: 1993
ADR: 3,III